Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TAF1B Antibody (OTI1A8), HRP, Novus Biologicals™
SDP

Catalog No. NBP274441H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP274441H 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP274441H Supplier Novus Biologicals Supplier No. NBP274441H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TAF1B Monoclonal specifically detects TAF1B in Human samples. It is validated for Western Blot.

Specifications

Antigen TAF1B
Applications Western Blot
Classification Monoclonal
Clone OTI1A8
Conjugate HRP
Dilution Western Blot
Formulation PBS with No Preservative
Gene Alias RAF1BTATA box binding protein (TBP)-associated factor, RNA polymerase I, B, 63kD, RAFI63MGC:9349, RNA polymerase I-specific TBP-associated factor 63 kDa, SL1SL1, 63kD subunit, TAFI63TBP-associated factor 1B, TATA box binding protein (TBP)-associated factor, RNA polymerase I, B, 63kDa, TATA box-binding protein-associated factor 1B, TATA box-binding protein-associated factor RNA polymerase I subunit B, Transcription initiation factor SL1/TIF-IB subunit B
Gene Symbols TAF1B
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 1-208 of human TAF1B (NP_005671) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 9014.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.