Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TAF1C Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB290528 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB290528 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB290528 Supplier Novus Biologicals Supplier No. NBP212779APC
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TAF1C Polyclonal antibody specifically detects TAF1C in Human samples. It is validated for Western Blot, Immunoprecipitation

Specifications

Antigen TAF1C
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate APC
Dilution Western Blot, Immunoprecipitation
Formulation PBS
Gene Alias MGC:39976, RNA polymerase I-specific TBP-associated factor 110 kDa, SL1, 110kD subunit, SL1TATA box binding protein (TBP)-associated factor, RNA polymerase I, C, 110kD, TAFI110TBP-associated factor 1C, TAFI95, TATA box binding protein (TBP)-associated factor, RNA polymerase I, C, 110kDa, TATA box-binding protein-associated factor 1C, TATA box-binding protein-associated factor RNA polymerase I subunit C, transcription factor SL1, Transcription initiation factor SL1/TIF-IB subunit C
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residue 819 and 869 of human TATA Box Binding Protein (TBP)-Associated Factor, RNA Polymerase I, C, 110kDa using the numbering given in entry NP_005670.3 (GeneID 9013).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9013
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.