Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TAF2 Antibody (OTI2A5), DyLight 680, Novus Biologicals™
SDP

Catalog No. NBP274442FR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP274442FR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP274442FR Supplier Novus Biologicals Supplier No. NBP274442FR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TAF2 Monoclonal specifically detects TAF2 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen TAF2
Applications Western Blot
Classification Monoclonal
Clone OTI2A5
Conjugate DyLight 680
Dilution Western Blot
Formulation PBS with 0.05% Sodium Azide
Gene Alias 150 kDa cofactor of initiator function, CIF150TAFII-150, cofactor of initiator function, 150kD subunit, TAF(II)150, TAF2 RNA polymerase II, TATA box binding protein (TBP)-associated factor, TAF2B, TAFII150RNA polymerase II TBP-associated factor subunit B, TATA box binding protein (TBP)-associated factor, RNA polymerase II, B, 150kD, TBP-associated factor 150 kDa, Transcription initiation factor TFIID 150 kDa subunit, transcription initiation factor TFIID subunit 2
Gene Symbols TAF2
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 782-1121 of human TAF2 (NP_003175) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 6873.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.