Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TAF4B Antibody, DyLight 594, Novus Biologicals™
SDP

Catalog No. NB150112 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB150112 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB150112 Supplier Novus Biologicals Supplier No. NBP298110DL594
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TAF4B Polyclonal specifically detects TAF4B in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen TAF4B
Applications Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 594
Dilution Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias TAF(II)105, TAF2C2, TAF4b RNA polymerase II, TATA box binding protein (TBP)-associated factor, 105kDa, TAFII105, TAFII-105, TATA box binding protein (TBP)-associated factor 4B, TATA box binding protein (TBP)-associated factor, RNA polymerase II, C2, 105kD, transcription initiation factor TFIID 105 kD subunit, transcription initiation factor TFIID 105 kDa subunit, transcription initiation factor TFIID subunit 4B
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human TAF4B fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6875
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.