Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TAF5 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP186971
Description
TAF5 Polyclonal specifically detects TAF5 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
TAF5 | |
Polyclonal | |
Western Blot 0.04-0.4 ug/ml, Immunohistochemistry 1:20 - 1:50, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
TAF(II)100, TAF2DTATA box binding protein (TBP)-associated factor 2D, TAF5 RNA polymerase II, TATA box binding protein (TBP)-associated factor, TAFII100TAFII-100, TATA box binding protein (TBP)-associated factor, RNA polymerase II, D, 100kD, transcription initiation factor TFIID 100 kD subunit, Transcription initiation factor TFIID 100 kDa subunit, transcription initiation factor TFIID subunit 5 | |
Rabbit | |
Affinity Purified | |
RUO | |
6877 | |
Human | |
IgG |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
TAF5 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:IVQEHLYIDIFDGMPRSKQQIDAMVGSLAGEAKREANKSKVFFGLLKEPEIEVPLDDEDEEGENEEGKPKKKKPKKDSIGSKSKKQDPNAPPQNRIPLPELKDSDKLDKIMNMKETTK | |
0.1 mL | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction