Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TAF5 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $646.00
Specifications
Antigen | TAF5 |
---|---|
Dilution | Western Blot 0.04-0.4 ug/ml, Immunohistochemistry 1:20 - 1:50, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:20 - 1:50 |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
TAF5 Polyclonal specifically detects TAF5 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
TAF5 | |
Polyclonal | |
Rabbit | |
Human | |
TAF(II)100, TAF2DTATA box binding protein (TBP)-associated factor 2D, TAF5 RNA polymerase II, TATA box binding protein (TBP)-associated factor, TAFII100TAFII-100, TATA box binding protein (TBP)-associated factor, RNA polymerase II, D, 100kD, transcription initiation factor TFIID 100 kD subunit, Transcription initiation factor TFIID 100 kDa subunit, transcription initiation factor TFIID subunit 5 | |
TAF5 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Western Blot 0.04-0.4 ug/ml, Immunohistochemistry 1:20 - 1:50, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
6877 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:IVQEHLYIDIFDGMPRSKQQIDAMVGSLAGEAKREANKSKVFFGLLKEPEIEVPLDDEDEEGENEEGKPKKKKPKKDSIGSKSKKQDPNAPPQNRIPLPELKDSDKLDKIMNMKETTK | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title