Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TAF6L Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $646.00
Specifications
Antigen | TAF6L |
---|---|
Dilution | Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:20 - 1:50 |
Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
TAF6L Polyclonal specifically detects TAF6L in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
TAF6L | |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
FLJ11136, p300/CBP-associated factor (PCAF)-associated factor 65, PAF65-alpha, PAF65AMGC4288, PCAF-associated factor 65 alpha, PCAF-associated factor 65-alpha, TAF6-like RNA polymerase II p300/CBP-associated factor-associated factor 65 kDasubunit 6L, TAF6-like RNA polymerase II, p300/CBP-associated factor (PCAF)-associatedfactor, 65 kD, TAF6-like RNA polymerase II, p300/CBP-associated factor (PCAF)-associatedfactor, 65kDa | |
TAF6L | |
IgG | |
Affinity Purified |
Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
Polyclonal | |
Rabbit | |
Human | |
Q9Y6J9 | |
10629 | |
This antibody was developed against a recombinant protein corresponding to amino acids: LCCHYGAVVGLHALGWKAVERVLYPHLSTYWTNLQAVLDDYSVSNAQVKADGHKVYGAILVAVERLLKMKAQAAEPNRGGP | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title