Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TAF7L Antibody, Novus Biologicals™
SDP

Catalog No. NBP184350 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP184350 0.1 mL
NB432672 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP184350 Supplier Novus Biologicals Supplier No. NBP184350
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TAF7L Polyclonal specifically detects TAF7L in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen TAF7L
Applications Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:10-1:20
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Alias 50kDa, CT40FLJ23157, RNA polymerase II TBP-associated factor subunit Q, TAF2QRNA polymerase II, Q, TAF7-like RNA polymerase II, TATA box binding protein (TBP)-associated factor, TATA box binding protein (TBP)-associated factor, RNA polymerase II, Q, TATA box-binding protein-associated factor 50 kDa, transcription initiation factor TFIID subunit 7-like
Gene Symbols TAF7L
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids:ESQDEVPDEVENQFILRLPLEHACTVRNLARSQSVKMKDKLKIDLLPDGRHAVVEVEDVPLAAKLVDLPCVIESLRTLDKKTFYKTADISQMLVCTADGDIHLSPEEPAASTDPNIVRKKERGREEKCVWKHGITPPLKNVRK
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54457
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.