Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TAF9 Antibody, mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB343557 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB343557 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB343557 Supplier Novus Biologicals Supplier No. NBP241266MFV610
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TAF9 Polyclonal antibody specifically detects TAF9 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen TAF9
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias EC 2.7.4.3, MGC:5067, MGC1603, RNA polymerase II TBP-associated factor subunit G, STAF31/32, TAF2G, TAF9, TAF9 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 32kDa, TAFII31, TAFII-31, TAFII32, TAFII-32, TAFIID32, transcription initiation factor TFIID 31 kD subunit, Transcription initiation factor TFIID 31 kDa subunit, Transcription initiation factor TFIID 32 kDa subunit, transcription initiation factor TFIID subunit 9
Host Species Rabbit
Immunogen Antibody was raised against a 17 amino acid peptide near the center of human TAF9. The immunogen is located within amino acids 110 - 160 of TAF9. Amino Acid Squence: LTAPNYRLKSLQKKAS
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 6880
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.