Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TAFA4/FAM19A4 Antibody (480103), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB303326 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB303326 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB303326 Supplier Novus Biologicals Supplier No. FAB5099S

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

TAFA4/FAM19A4 Monoclonal antibody specifically detects TAFA4/FAM19A4 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF

Specifications

Antigen TAFA4/FAM19A4
Applications Western Blot, Flow Cytometry, CyTOF
Classification Monoclonal
Clone 480103
Conjugate Alexa Fluor 750
Dilution Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias Chemokine-like protein TAFA-4, family with sequence similarity 19 (chemokine (C-C motif)-like), member A4, FLJ25161, TAFA4, TAFA-4
Host Species Mouse
Immunogen E. coli-derived recombinant human TAFA4, Ser35-Arg140, Accession # Q96LR4
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 151647
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.