Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TAFA4/FAM19A4 Antibody (480103), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB347456 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB347456 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB347456 Supplier Novus Biologicals Supplier No. FAB5099MFV610
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TAFA4/FAM19A4 Monoclonal specifically detects TAFA4/FAM19A4 in Human samples. It is validated for CyTOF, Flow Cytometry (Intracellular Staining), Western Blot.

Specifications

Antigen TAFA4/FAM19A4
Applications CyTOF, Flow Cytometry (Intracellular Staining), Western Blot
Classification Monoclonal
Clone 480103
Conjugate mFluor Violet 610 SE
Dilution Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias Chemokine-like protein TAFA-4, family with sequence similarity 19 (chemokine (C-C motif)-like), member A4, FLJ25161, TAFA4, TAFA-4
Host Species Mouse
Immunogen E. coli-derived recombinant human TAFA4, Ser35-Arg140, Accession # Q96LR4
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 151647
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.