Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tafazzin/TAZ Antibody, Novus Biologicals™
SDP

Catalog No. NB600811 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB600811 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB600811 Supplier Novus Biologicals Supplier No. NB600811
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Tafazzin/TAZ Polyclonal specifically detects Tafazzin/TAZ in Human samples. It is validated for Western Blot, Peptide ELISA.

Specifications

Antigen Tafazzin/TAZ
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.3 - 10 ug/ml, Peptide ELISA Detection limit 1:32000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_000107.1
Gene Alias BTHSEFE, cardiomyopathy, dilated 3A (X-linked), CMD3A, EFE2FLJ27390, G4.5endocardial fibroelastosis 2, LVNCX, Protein G4.5, tafazzin, Taz1, XAP-2
Gene Symbols TAFAZZIN
Host Species Goat
Immunogen Peptide with sequence C-HLKTQAEQLHNH corresponding to C-Terminus according to NP_000107.1, NP_851828.1, NP_851829.1, NP_851830.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 6901
Test Specificity This antibody is expected to recognize isoforms 1to 4 (NP_000107.1; NP_851828.1; NP_851829.1; NP_851830.1, respectively).
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.