Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TAG-72 Antibody (CA72/2869R), DyLight 755, Novus Biologicals™
SDP

Catalog No. NB385247 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB385247 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB385247 Supplier Novus Biologicals Supplier No. NBP308661IR
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

TAG-72 Monoclonal antibody specifically detects TAG-72 in Human, Rat, Bovine, Canine, Hamster samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen TAG-72
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CA72/2869R
Conjugate DyLight 755
Formulation 50mM Sodium Borate
Gene Alias CA 72-4, Protein TAG-72, tag 72, tag-72 Protein TAG-72
Host Species Rabbit
Immunogen Purified TAG-72 protein
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Tumor Biomarkers
Primary or Secondary Primary
Target Species Human, Rat, Bovine, Canine, Hamster
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.