Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TAGAP Antibody (SR2094), Novus Biologicals™
SDP

Catalog No. NB258060 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB258060 50 μg
NB258100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB258060 Supplier Novus Biologicals Supplier No. NBP32260350UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

TAGAP Monoclonal antibody specifically detects TAGAP in Human, Mouse, Rat samples. It is validated for Western Blot, Immunoprecipitation

Specifications

Antigen TAGAP
Applications Western Blot, Immunoprecipitation
Classification Monoclonal
Clone SR2094
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Immunoprecipitation 1:50
Formulation PBS, pH 7.4, 150mM NaCl, 50% glycerol.
Gene Alias FLJ32631, FLJ39771, IDDM21, MGC133247, MGC27381, TAGAP1, T-cell activation GTPase activating protein, T-cell activation GTPase-activating protein, T-cell activation Rho GTPase-activating protein, T-cell activation RhoGTPase activating protein
Host Species Rabbit
Immunogen A synthesized peptide derived from human TAGAP (Uniprot #: Q8N103)
Purification Method Affinity purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 117289
Target Species Human, Mouse, Rat
Content And Storage Store at -20° C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.