Learn More
Tocris Bioscience™ TAK 165

Description
Specifications
Specifications
| Quantity | 10 mg |
| Product Type | TAK 165 |
| Purity | >99% |
| Molecular Weight (g/mol) | 468.47 |
| Inhibitors | TAK 165 |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.