Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ TAK 715
GREENER_CHOICESDP

Catalog No. 425410 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
42-541-0 10 mg
42-545-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 42-541-0 Supplier Tocris Bioscience™ Supplier No. 4254/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent p38 MAPK inhibitor; anti-inflammatory

Inhibitor of p38 MAPK (IC50 = 7.1 nM for p38 MAPKα). Also inhibits Wnt-3a-stimulated β-catenin signaling. Inhibits 22 kinases by more than 80%, including CK1δ/ε. Anti-inflammatory and anti-rheumatoid arthritis agent. Orally bioavailable.

Chemical Identifiers

CAS 303162-79-0
Molecular Formula C24H21N3OS

Specifications

Quantity 10 mg
Formula Weight 399.51
Purity >99%
Molecular Weight (g/mol) 399.512
Chemical Name or Material TAK 715
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.