Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TAK1 Antibody, Novus Biologicals™
SDP

Catalog No. NB10056363 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056363 0.1 mg
NB10056363T 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10056363 Supplier Novus Biologicals Supplier No. NB10056363
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 2 publications

TAK1 Polyclonal specifically detects TAK1 in Human, Mouse samples. It is validated for IF/IHC, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen TAK1
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-2 ug/ml, Immunohistochemistry 1:200. Use reported in scientific literature (PMID 30786091), Immunocytochemistry/Immunofluorescence 2-5 ug/ml, Immunohistochemistry-Paraffin 1:200
Formulation PBS containing 0.05% BSA with 0.05% Sodium Azide
Gene Accession No. O43187
Gene Alias EC 2.7.11, EC 2.7.11.25, MEKK7, mitogen-activated protein kinase kinase kinase 7, TAK1/MAP3K7, TAK1TGF1a, TGF-beta activated kinase 1, TGF-beta-activated kinase 1, Transforming growth factor-beta-activated kinase 1
Gene Symbols MAP3K7
Host Species Rabbit
Immunogen This polyclonal antibody was raised against amino acids 19-35 and 564-579 of human TAK1 (accession number BAA25025).
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Apoptosis, Immunology, Innate Immunity, Protein Kinase, Signal Transduction, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 6885
Test Specificity In NIH 3T3, an approx. 70 kDa band is observed.
Target Species Human, Mouse, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.