Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TAK1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 2 publications
$206.50 - $427.50
Specifications
Antigen | TAK1 |
---|---|
Concentration | 0.5 mg/mL |
Dilution | Western Blot 1-2 ug/ml, Immunohistochemistry 1:200. Use reported in scientific literature (PMID 30786091), Immunocytochemistry/Immunofluorescence 2-5 ug/ml, Immunohistochemistry-Paraffin 1:200 |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB10056363T
![]() |
Novus Biologicals
NB10056363SS |
0.025 mg |
Each for $206.50
|
|
|||||
NB10056363
![]() |
Novus Biologicals
NB10056363 |
0.1 mg |
Each for $427.50
|
|
|||||
Description
TAK1 Polyclonal specifically detects TAK1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
TAK1 | |
Western Blot 1-2 ug/ml, Immunohistochemistry 1:200. Use reported in scientific literature (PMID 30786091), Immunocytochemistry/Immunofluorescence 2-5 ug/ml, Immunohistochemistry-Paraffin 1:200 | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse | |
O43187 | |
6885 | |
This polyclonal antibody was raised against amino acids 19-35 and 564-579 of human TAK1 (accession number BAA25025). | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
0.5 mg/mL | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Apoptosis, Immunology, Innate Immunity, Protein Kinase, Signal Transduction, Wnt Signaling Pathway | |
PBS containing 0.05% BSA with 0.05% Sodium Azide | |
EC 2.7.11, EC 2.7.11.25, MEKK7, mitogen-activated protein kinase kinase kinase 7, TAK1/MAP3K7, TAK1TGF1a, TGF-beta activated kinase 1, TGF-beta-activated kinase 1, Transforming growth factor-beta-activated kinase 1 | |
MAP3K7 | |
IgG | |
Protein G purified | |
In NIH 3T3, an approx. 70 kDa band is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title