Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TAK1 Antibody, Biotin, Novus Biologicals™
SDP

Catalog No. NB206724 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB206724 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB206724 Supplier Novus Biologicals Supplier No. NBP176441B
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TAK1 Polyclonal antibody specifically detects TAK1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA

Specifications

Antigen TAK1
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Biotin
Formulation PBS
Gene Alias EC 2.7.11, EC 2.7.11.25, MEKK7, mitogen-activated protein kinase kinase kinase 7, TAK1/MAP3K7, TAK1TGF1a, TGF-beta activated kinase 1, TGF-beta-activated kinase 1, Transforming growth factor-beta-activated kinase 1
Host Species Rabbit
Immunogen Antibody was raised against a synthetic peptide corresponding to 14 amino acids near the amino-terminus of human TAK1 isoform a. The immunogen is located within amino acids 40 - 90 of TAK1. Amino Acid Squence: AIKQIESESERKAF
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Immunology, Innate Immunity, Protein Kinase, Signal Transduction, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 6885
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.