Learn More
Eisco™ Talc Mineral Specimen
Click to view available options
Quantity:
1 Ea.
10/Pk.
12/Pk.
Description
- Hot fluid infiltrates atomic structure of other silicate minerals by metasomatism, altering them to form talc
- For use in learning properties of minerals that have been thermally altered
- For use in classroom identification labs and exercises
- Correlates well to Next Generation Science Standards Evidence Statements
- For use in study of mineral hardness as its softness contrasts well with harder mineral such as quartz
Specifications
Specifications
Product Type | Mineral, Talc |
For Use With (Application) | Lessons in mineral hardness |
Packaging | Clear labeled and barcoded bag |
Quantity | 1 Ea. |
Specimen Size | 2 to 3 cm |
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov