Learn More
Eisco™ Talc Mineral Specimen
SCI_ED
Catalog No. S26593
Click to view available options
Quantity:
1 Ea.
10/Pk.
12/Pk.
Talc Mineral Specimen is at bottom of Mohs scale of hardness, and has distinct soapy feel.
  • Hot fluid infiltrates atomic structure of other silicate minerals by metasomatism, altering them to form talc
  • For use in learning properties of minerals that have been thermally altered
  • For use in classroom identification labs and exercises
  • Correlates well to Next Generation Science Standards Evidence Statements
  • For use in study of mineral hardness as its softness contrasts well with harder mineral such as quartz
Specifications
Product Type Mineral, Talc
Quantity 10/Pk.
Specimen Size 2 to 3 cm
Weight (Metric) 0.2 kg
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.