Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Tali™ Image-Based Cytometer Power Cord

Catalog No. T10793
Encompass_Preferred
Change view
Click to view available options
Quantity:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T10793 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T10793 Supplier Invitrogen™ Supplier No. T10793
Only null left
Add to Cart
Add to Cart

This power cord is a replacement part for the Tali™ Image-Based Cytometer instrument.

This power cord is a replacement part for the Tali™ Image-Based Cytometer instrument. The Tali™ Image-Based Cytometer Power cord and the four universal adaptors included have been specifically designed for use with the Tali™ Image-Based Cytometer instrument. This product has been CE certified.

TRUSTED_SUSTAINABILITY

Specifications

Type Cytometer Power Cord
Product Line Tali
For Use With (Equipment) Tali™ Image-Based Cytometer
Content And Storage 1 replacement power cord for the Tali™ Image Based Cytometer.

Includes 4 adapter plugs (for U.S./E.U./U.K./Australia).
Quantity Each
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.