Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Talin1 Antibody (97H6), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB309585 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB309585 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB309585 Supplier Novus Biologicals Supplier No. NBP250320MFV610
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Talin1 Monoclonal antibody specifically detects Talin1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation

Specifications

Antigen Talin1
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunoprecipitation
Classification Monoclonal
Clone 97H6
Conjugate mFluor Violet 610 SE
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation
Formulation 50mM Sodium Borate
Gene Alias ILWEQ, KIAA1027, talin 1, talin-1, TLN
Host Species Mouse
Immunogen His-tagged polypeptide containing residues 489-911 of mouse Talin1
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers
Primary or Secondary Primary
Gene ID (Entrez) 7094
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.