Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Talin1 Antibody (TA205), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NBP121642W Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP121642W 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP121642W Supplier Novus Biologicals Supplier No. NBP121642AF405
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Talin1 Monoclonal specifically detects Talin1 in Human, Chicken, Rabbit samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.

Specifications

Antigen Talin1
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone TA205
Conjugate Alexa Fluor 405
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen
Gene Accession No. Q9Y490
Gene Alias ILWEQ, KIAA1027, talin 1, talin-1, TLN
Gene Symbols TLN1
Host Species Mouse
Immunogen Human platelet talin.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cytoskeleton Markers
Primary or Secondary Primary
Gene ID (Entrez) 7094
Test Specificity Immunoblots the 225 kDa talin band on SDS gels of platelets and other cells. It may be used to stain adhesion plaques in fixed and permeabilized fibroblasts. TA205 recognizes an epitope within residues 139-433.
Target Species Human, Chicken, Rabbit
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.