Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Talin1, p Ser425 Antibody, Novus Biologicals™
SDP

Catalog No. NB387824 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB387824 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB387824 Supplier Novus Biologicals Supplier No. NBP323437
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Talin1 Polyclonal antibody specifically detects Talin1 in Human, Mouse, Rat, Chicken, Xenopus samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen Talin1
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunocytochemistry/ Immunofluorescence 1:200
Formulation PBS, 1 mg/mL BSA and 50% Glycerol
Gene Alias ILWEQ, KIAA1027, talin 1, talin-1, TLN
Host Species Rabbit
Immunogen Phospho-Talin1 (Ser-425) synthetic peptide (coupled to carrier protein) corresponding to amino acids surrounding Ser-425 in human Talin1. This sequence is conserved in rat, mouse, and chicken Talin1, as well as in Talin2 (Ser-428). [UniProt# Q9Y490]
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers
Primary or Secondary Primary
Gene ID (Entrez) 7094
Target Species Human, Mouse, Rat, Chicken, Xenopus species
Content And Storage Store at -20°C.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.