Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva TALON™ Superflow Medium

Catalog No. p-4415995
Click to view available options
Quantity:
10 mL
50 mL

Offers different selectivity compared to nickel-charged media. Cytiva TALON™ Superflow Medium used for purification of histidine-tagged recombinant proteins by IMAC.

Features:

  • Ligand - TALON Superflow Cobalt-IMAC
  • Matrix - Cross-linked agarose, 6%
  • Particle Size - 60 -160μm
  • Binding Capacity/mL - Up to 20 mg histidine-tagged protein
  • pH stability working range - 3 to 12
  • pH stability Cleaning-in-Place (CIP) - 2 - 14 Co2+stripped medium
  • Complete packsize - 10mL

 

TRUSTED_SUSTAINABILITY

Specifications

Quantity 10 mL
For Use With (Application) For PurifiCatIon of Histidine-Tagged Recombinant Pby Immobilized Metal Affinity Chromatography
Storage Requirements 4°C to 8°C
Binding Capacity Up to 20 mg Histidine-tagged Protein
Product Type Talon Superflow Media
Particle Size 60 to 160 μm
Matrix 6% Cross-Linked Agarose
pH Range 3 to 12
Storage Buffer 20% Ethanol
Ligand Type Talon Superflow Cobalt-IMAC
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.