Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TANC1 Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB291221 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB291221 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB291221 Supplier Novus Biologicals Supplier No. NBP152630APC
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TANC1 Polyclonal antibody specifically detects TANC1 in Human samples. It is validated for Western Blot, Immunoprecipitation

Specifications

Antigen TANC1
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate APC
Dilution Western Blot, Immunoprecipitation
Formulation PBS
Gene Alias FLJ13360, FLJ41166, KIAA1728, protein TANC1, rolling pebbles homolog B, ROLSB, TANC, tetratricopeptide repeat, ankyrin repeat and coiled-coil containing 1, tetratricopeptide repeat, ankyrin repeat and coiled-coil domain-containing protein 1, TPR domain, ankyrin-repeat and coiled-coil-containing
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 1 and 50 of human Tetratricopeptide Repeat, Ankyrin Repeat and Coiled-Coil Containing 1 using the numbering given in entry NP_203752.2 (GeneID 85461).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 85461
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.