Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Tandospirone hydrochloride
GSA_VAGREENER_CHOICE

Catalog No. 285410
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective 5-HT1A partial agonist

5-HT1A receptor partial agonist (Ki = 27 nM) that displays selectivity over 5-HT2, 5-HT1C, α1, α2, D1 and D2 receptors (Ki values ranging from 1300 - 41000 nM). Inactive at 5-HT uptake sites, 5-HT1B, β-adrenergic, muscarinic and benzodiazepine receptors. Displays anxiolytic activity.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Chemical Identifiers

CAS 99095-10-0
Molecular Formula C21H30ClN5O2

Specifications

Quantity 10 mg
Formula Weight 419.95
Purity >98%
Molecular Weight (g/mol) 419.95
Chemical Name or Material Tandospirone hydrochloride
Recommended Storage Store at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.