Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Tango™ EDG7-bla U2OS Cells

Catalog No. K1849
Encompass_Preferred
Change view
Click to view available options
Quantity:
8 x 106 cells/vial
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
K1849 8 x 106 cells/vial
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. K1849 Supplier Thermo Scientific™ Supplier No. K1849

Please call customer service for current pricing. 1-800-766-7000

Tango™ EDG7-bla U2OS cells contain the human Endothelial Differentiation Gene 7 (EDG7) linked to a TEV protease site and a Gal4-VP16 transcription factor stably integrated into the Tango™ GPCR-bla U2OS parental cell line.

Tango™ EDG7-bla U2OS cells contain the human Endothelial Differentiation Gene 7 (EDG7) linked to a TEV protease site and a Gal4-VP16 transcription factor stably integrated into the Tango™ GPCR-bla U2OS parental cell line. This parental cell line stably expresses a beta-arrestin/TEV protease fusion protein and the beta-lactamase reporter gene under the control of a UAS response element. The Tango™ EDG7-bla U2OS cells have been functionally validated for Z' factor and EC50 concentrations of LPA. In addition, Tango™ EDG7-bla U2OS cells have been tested for assay performance under variable conditions.

TRUSTED_SUSTAINABILITY

Specifications

Accession Number NM_012152.1
Assay Entry Cell-based beta-arrestin recruitment
Cell State Dividing Cells
Content And Storage Tango™ cell lines are shipped on dry ice. Store in liquid nitrogen immediately upon receipt, or thaw for immediate use.
Cytoplasmic Tail Native Tail
Detection Method Fluorescence
For Use With (Application) FRET
Human Protein Class G-Protein Coupled Receptor (GPCR)
Population Status Single Cell Clone Origin
Product Line Tango
Quantity 8 x 106 cells/vial
Readout End Point
Reporter Gene BLA (Beta-Lactamase)
Target Entry EDG7
Cell Line U2OS
System Type Tango™
Druggable Target GPCR
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.