Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Tango™ OPRL1-bla U2OS Cells

Catalog No. K1786
Encompass_Preferred
Change view
Click to view available options
Quantity:
8 x 106 Cells
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
K1786 8 x 106 Cells
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. K1786 Supplier Thermo Scientific™ Supplier No. K1786

Please call customer service for current pricing. 1-800-766-7000

The Tango™ OPRL1-bla U2OS cells contain the human Opioid Receptor-Like 1 (OPRL1) linked to a TEV protease site and a Gal4-VP16 transcription factor stably integrated into the Tango™ GPCR-bla U2OS parental cell line.

The Tango™ OPRL1-bla U2OS cells contain the human Opioid Receptor-Like 1 (OPRL1) linked to a TEV protease site and a Gal4-VP16 transcription factor stably integrated into the Tango™ GPCR-bla U2OS parental cell line. This parental cell line stably expresses a beta-arrestin/TEV protease fusion protein and the beta-lactamase (bla) reporter gene under the control of a UAS response element.

The Tango™ OPRL1-bla U2OS cells have been functionally validated for Z'-factor and EC50 concentrations of established ligands. In addition, Tango™ OPRL1-bla U2OS cells have been tested for assay performance under variable conditions. These data are found in the Validation & Assay Performance Summary for each product.

TRUSTED_SUSTAINABILITY

Specifications

Accession Number NM_000913
Assay Entry Cell-based beta-arrestin recruitment
Cell State Dividing Cells
Content And Storage LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol.

Includes:
• Tango™ OPRL1-bla U2OS cells
• Tango™ OPRL1-bla U2OS Assay Protocol
• Certificate of Analysis

The Tango™ OPRL1-bla U2OS cells are shipped on dry ice and should be stored in liquid nitrogen immediately upon receipt.
Cytoplasmic Tail Tail from AVPR2 GPCR
Detection Method Fluorescence
For Use With (Application) FRET
Gene Symbol OPRL1
Human Protein Class G-Protein Coupled Receptor (GPCR)
Population Status Single Cell Clone Origin
Product Line GeneBLAzer
Protein Family GPCR (G-Protein Coupled Receptors)
Quantity 8 x 106 Cells
Readout End Point
Reporter Gene BLA (Beta-Lactamase)
Target Entry OPRL1
Type U2OS Cell
Cell Line U2OS
System Type Tango™
Druggable Target GPCR
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.