Learn More
Invitrogen™ Tango™ OPRM1-bla U2OS Cells
Description
The Tango™ OPRM1-bla U2OS cells have been functionally validated for Z' factor and EC50 concentrations of DAMGO. In addition, Tango™ OPRM1-bla U2OS cells have been tested for assay performance under variable conditions. These data are found in the Validation & Assay Performance Summary in the Manuals and Brochures section.
Specifications
Specifications
| Accession Number | NM_000914 |
| Assay Entry | Cell-based beta-arrestin recruitment |
| Cell State | Dividing Cells |
| Content And Storage | (LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol). Includes: o Tango™ OPRM1-bla U2OS cells o Tango™ OPRM1-bla U2OS Assay Protocol o Certificate of Analysis The Tango™ OPRM1-bla U2OS cells are shipped on dry ice and should be stored in liquid nitrogen immediately upon receipt. |
| Cytoplasmic Tail | Tail from AVPR2 GPCR |
| Detection Method | Fluorescence |
| Gene Symbol | OPRM1 |
| Population Status | Single Cell Clone Origin |
| Product Line | Tango |
| Quantity | 2 x 106 Cells |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.