Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TANGO2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $499.50
Specifications
Antigen | C22orf25 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP17046320
![]() |
Novus Biologicals
NBP17046320UL |
20 μL |
Each for $206.00
|
|
|||||
NBP170463
![]() |
Novus Biologicals
NBP170463 |
100 μL |
Each for $499.50
|
|
|||||
Description
TANGO2 Polyclonal specifically detects TANGO2 in Human samples. It is validated for Western Blot.Specifications
C22orf25 | |
Polyclonal | |
Rabbit | |
chromosome 22 open reading frame 25 | |
TANGO2 | |
IgG | |
31 kDa |
Western Blot | |
Unconjugated | |
RUO | |
128989 | |
Synthetic peptides corresponding to C22ORF25 The peptide sequence was selected from the N terminal of C22ORF25. Peptide sequence MCIIFFKFDPRPVSKNAYRLILAANRDEFYSRPSKLADFWGNNNEILSGL. | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title