Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TAO Kinase 1 Antibody (TAO1 1.2), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NP20324A32 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP20324A32 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP20324A32 Supplier Novus Biologicals Supplier No. NBP250324AF532
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TAO Kinase 1 Monoclonal specifically detects TAO Kinase 1 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Knockdown Validated.

Specifications

Antigen TAO Kinase 1
Applications Western Blot, Immunocytochemistry, Immunofluorescence, KnockDown
Classification Monoclonal
Clone TAO1 1.2
Conjugate Alexa Fluor 532
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias EC 2.7.11, FLJ14314, hKFC-B, KIAA1361STE20-like kinase PSK2, Kinase from chicken homolog B, MAP3K16EC 2.7.11.1, MARKKmicrotubule affinity regulating kinase kinase, PSK2serine/threonine kinase TAO1, serine/threonine-protein kinase TAO1, TAO kinase 1, TAO1serine/threonine protein kinase TAO1 homolog, Thousand and one amino acid protein 1
Gene Symbols TAOK1
Host Species Mouse
Immunogen His-tagged recombinant human TAO Kinase 1 (722-1001) generated in E.coli
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline GPCR, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 57551
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.