Learn More
TAOK1 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$167.95
Specifications
| Antigen | TAOK1 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Liquid |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-155-7834
|
Bethyl Laboratories, Inc
A300524AT |
20 μL |
Each of 1 for $167.95
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse
TAOK1 is a TAO type protein kinase. TAOK1 expression has been documented in rat brain. ESTs have been isolated from human normal adrenal, bone and breast and cancerous duodenum, cervix and germ cell.Specifications
| TAOK1 | |
| Polyclonal | |
| Liquid | |
| RUO | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| FLJ14314, hKFC-B, hTAOK1, KFC-B, KIAA1361, kinase from chicken homolog B, MAP3K16, MARK Kinase, MARKK, microtubule affinity regulating kinase kinase, prostate-derived STE20-like kinase 2, prostate-derived sterile 20-like kinase 2, PSK2, PSK-2, serine/threonine protein kinase TAO1 homolog, STE20-like kinase PSK2, TAO1, TAOK1, thousand and one amino acid protein 1, thousand and one amino acid protein kinase 1 | |
| TAOK1 | |
| IgG | |
| Antigen affinity chromatography | |
| Antibody |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q7L7X3 | |
| 57551 | |
| Between 400 and 450 | |
| Primary | |
| 4° C | |
| TAOK1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.