Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tapasin Antibody, Novus Biologicals™
SDP

Catalog No. NBP237749 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP237749 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP237749 Supplier Novus Biologicals Supplier No. NBP237749
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Tapasin Polyclonal specifically detects Tapasin in Human samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen Tapasin
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunoprecipitation 1:100
Formulation PBS and 50% Glycerol with 0.09% Sodium Azide
Gene Alias NGS17, NGS-17, TAP binding protein (tapasin), tapasin, TAPATAP-associated protein, TPN, TPSNTAP-binding protein
Gene Symbols TAPBP
Host Species Rabbit
Immunogen Synthetic peptide derived from the sequence of human Tapasin
Purification Method Protein A purified
Quantity 0.05 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6892
Test Specificity Tapasin
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.