Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tapasin Related Protein Antibody (OTI4H5), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. N274443J549 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N274443J549 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N274443J549 Supplier Novus Biologicals Supplier No. NBP274443JF549
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Tapasin Related Protein Monoclonal specifically detects Tapasin Related Protein in Human, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Tapasin Related Protein
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI4H5
Conjugate Janelia Fluor 549
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias FLJ10143, TAP binding protein-like, Tapasin-like, TAPASIN-R, tapasin-related protein, TAP-binding protein-like, TAP-binding protein-related protein, TAPBP-RTAP binding protein related, TAPBPRtapasin-like
Gene Symbols TAPBPL
Host Species Mouse
Immunogen Full length recombinant protein of human Tapasin Related Protein (NP_060479) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 55080.0
Target Species Human, Monkey
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.