Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TAPBPR Antibody (1059329), DyLight 405, Novus Biologicals™
SDP

Catalog No. NB305258 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB305258 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB305258 Supplier Novus Biologicals Supplier No. FAB11337E
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TAPBPR Monoclonal antibody specifically detects TAPBPR in Human samples. It is validated for Flow Cytometry

Specifications

Antigen TAPBPR
Applications Flow Cytometry
Classification Monoclonal
Clone 1059329
Conjugate DyLight 405
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias FLJ10143, TAP binding protein-like, Tapasin-like, TAPASIN-R, tapasin-related protein, TAP-binding protein-like, TAP-binding protein-related protein, TAPBP-RTAP binding protein related, TAPBPRtapasin-like
Host Species Mouse
Immunogen Chinese Hamster Ovary cell line, CHO-derived human TAPBPR, Ala19-Arg404, Accession # Q9BX59.2
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55080
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.