Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MP Biomedicals™ Taq Core Kit 10
Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
10 x 250U
1000U
5 x 1000U
5 x 250U
Description
Includes
Taq DNA Polymerase (5U/μL), 10X Standard Buffer with Mg, 10X Standard Buffer without Mg, 25mM MgCl2, 10mM dNTPs mix
Unit definition: One unit of DNA polymerase is the amount of enzyme required to catalyse the incorporation of 10nmol of nucleosides into a DE81 adsorbable product within 30 min. at 74°C under assay conditions.
Unit definition: One unit of DNA polymerase is the amount of enzyme required to catalyse the incorporation of 10nmol of nucleosides into a DE81 adsorbable product within 30 min. at 74°C under assay conditions.

Specifications
Specifications
Content And Storage | Taq DNA Polymerase (5U/μL), 10X Standard Buffer with Mg, 10X Standard Buffer without Mg, 25mM MgCl2, 10mM dNTPs mix. Store at -20°C |
Form | Frozen |
Format | Frozen liquid |
Quantity | 1000U |
For Use With (Application) | Standard PCR |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Research Use Only (RUO)
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction