Learn More
MP Biomedicals™ Taq Core Kit 25
Taq DNA Polymerase + dNTPs in one kit
$217.00
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MP1EPTQK251
View Documents
MP Biomedicals™
11EPTQK251
250 u
Each for $217.00
Only null left
Add to Cart
MP1EPTQK253
View Documents
MP Biomedicals™
11EPTQK253
3 x 250 u N/A N/A N/A
Description
Description
  • Taq CORE Kits contain all the reagents (presented as separate items) required for PCR:
  • MP Biomedicals high quality recombinant Taq DNA polymerase, high purity dNTPs Mix (>99%), optimized incubation buffers with and without MgCl2 and a separate MgCl2 solution.
  • dNTPs at 25mM each
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.