Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Taq DNA Polymerase (cloned)
GSA_VA

For PCR and other procedures requiring DNA polymerase activity at elevated temperatures

$184.12 - $996.03

Specifications

Concentration 5000 U/mL
Content And Storage 100mM Tris HCl, pH 9, 15mM MgCl2 and 500mM KCl. Store at -20°C
pH 7.5
Purity or Quality Grade Free from detectable nonspecific nuclease and Taq I restriction endonuclease activities.
For Use With (Application) Standard PCR
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
46-001-008
GSA_VA
View Documents
Cytiva
27079804
250U
Each for $184.12
Only null left
Add to Cart
 
46-001-010
View Documents
Cytiva
27079806
10 x 250U
Each for $996.03
Only null left
Add to Cart
 
Description

Description

One unit incorporates 10nmol of total nucleotides into acid-insoluble material in 30 min. at 70°C in a total volume of 50μL.

  • Purity: Free from detectable nonspecific nuclease and Taq I restriction endonuclease activities
  • Concentration: 5000 units/mL
  • Functional testing: Tested in PCR
  • Storgae Buffer: 50mM Tris-HCl (pH 7.5), 5mM DTT, 0.1mM EDTA, 50% glycerol, stabilizers

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

5000 U/mL
7.5
Standard PCR
100mM Tris HCl, pH 9, 15mM MgCl2 and 500mM KCl. Store at -20°C
Free from detectable nonspecific nuclease and Taq I restriction endonuclease activities.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.