Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Taq-&LOAD™ PCR Mastermix

This mastermix allows direct loading after the PCR on the agarose gel without the need of loading dye.

$322.50 - $2410.00

Specifications

Concentration 5X
Content And Storage Recombinant Taq DNA Polymerase, dNTPs, MgCl2, incubation and loading buffer. Store at -20°C
Format Tube
Gel Compatibility Agarose
Label or Dye Red-purple
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
11EPTAL100
View Documents
MP Biomedicals™
11EPTAL100
100U
Each for $322.50
Only null left
Add to Cart
 
11EPTAL110
View Documents
MP Biomedicals™
11EPTAL110
10 x 100U
Each for $2,410.00
Only null left
Add to Cart
 
Description

Description

Taq-&LOAD PCR Mastermix is a 5X concentrated solution containing MP Biomedicals recombinant Taq DNA Polymerase, dNTPs, MgCl2, incubation and loading buffer (includes a compound increasing the density of the sample and a red/purple dye). The included red/purple dye migrates at 400 bp on a 1.2% agarose gel.

Specifications

Specifications

5X
Tube
Red-purple
Recombinant Taq DNA Polymerase, dNTPs, MgCl2, incubation and loading buffer. Store at -20°C
Agarose
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.