Learn More
Applied Biosystems™ TaqMan™ Environmental Master Mix 2.0
Description
Applied Biosystems™ TaqMan™ Environmental Master Mix 2.0 offers accurate, real-time PCR-based pathogen detection in the presence of high levels of inhibitors. Use this Master Mix to analyze environmental, food and other challenging samples.
- Tailored for quantitative, real-time PCR experiments
- Unrivalled sensitivity for challenging applications, low copy number target detection and multiplex PCR
- Includes pathogen detection and viral⁄bacterial load quantification
- Sensitive detection for reliable quantification of abundant and limited genetic targets
- Stable mix for high-throughput handling
- Upon receipt, store at -15°C to -25°C before first use, protected from light
- After first use, store at 2°C to 8°C, protected from light
Optimized Formulation for Unrivalled Performance
TaqMan Environmental Master Mix 2.0 is a convenient 2X mix for target quantification that includes:
- AmpliTaq Gold™ DNA Polymerase, UP (Ultra Pure), a highly purified DNA Polymerase for improved detection of bacterial targets. This hot-start enzyme is inactive at room temperature so reactions can be set up on the benchtop. Enzyme is activated during thermal cycling.
- Passive internal reference based on proprietary ROX™ dye for precise data analysis.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Specifications
| Concentration | 2X |
| Content And Storage | This TaqMan™ Environmental Master Mix 2.0 is a convenient 2X mix in a 10 mL bottle that includes: AmpliTaq Gold DNA Polymerase, UP (Ultra Pure), and a passive internal reference based on proprietary ROX dye for precise data analysis. Upon receipt, store at -15 to -25°C before first use, protected from light. After first use, store at 2 to 8°C, protected from light. |
| Detection Method | Primer-probe |
| Format | Bottle |
| PCR Method | qPCR |
| Polymerase | AmpliTaq Gold DNA Polymerase |
| Reaction Speed | Standard |
| For Use With (Equipment) | 7000 System, 7300 System, 7500 Fast System, 7500 System, 7900HT Fast System, 7900HT System, StepOne™, Standard Mode |
| Passive Reference Dye | ROX (Pre-mixed) |
| Product Line | TaqMan |
| Show More |
Frequently Asked Questions (FAQs)
No, the 2X Environmental Master Mix (EMM) is not the same as the TaqMan Universal PCR Master Mix (with or without AmpErase UNG). The 2X Environmental Master Mix is specifically formulated to detect specific bacterial pathogens with greater specificity and sensitivity. It includes AmpliTaq Gold DNA Polymerase, LD, which is a highly purified version of AmpliTaq Gold DNA Polymerase. Every effort has been made in the development of this new polymerase to eliminate bacterial DNA from the enzyme preparation.
Yes, it is.
Yes. You can purchase the TaqMan Environmental Master Mix 2.0 (Cat. No. 4396838).
For fecal samples, we recommend a master mix that is very tolerant of inhibitors, such as our TaqMan Environmental Master Mix 2.0. Please refer to the following paper: https://assets.thermofisher.com/TFS-Assets/CMD/posters/cms_089094.pdf
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.