Learn More
Applied Biosystems™ TaqMan™ Genotyping Master Mix
Description
- Distinct clusters and high call rates for unambiguous allelic discrimination
- Validated with TaqMan™ SNP Genotyping Assays
- Excellent pre- and post-PCR stability for high throughput setup and analysis
- Reaction size is based on a 50μL reaction volume
May be used for a variety of genotyping applications, including:
- Candidate gene studies
- Drug-target validation
- Disease association studies
- Population genetics
- Linkage mapping
- Agricultural applications
Exceptional Performance from an Optimized Mix (2X)
Accurate genotype assignments result from preferential binding of the allele-specific probe to the matching target. TaqMan™ Genotyping Master Mix enables specific binding of the probe to achieve exceptional cluster resolution. Key features include:
- AmpliTaq Gold™ DNA Polymerase, UP (Ultra Pure) - Automatic hot start enzyme designed to be active during thermal cycling and inactive at room temperature for easy reaction setup
- Optimized mix components provide excellent specificity for discrimination between alleles
- Passive internal reference based on proprietary ROX™ dye for increased precision on Applied Biosystems™ real-time PCR instruments
- Single thermal cycling condition for consistent results with TaqMan™ Assays
Validated with TaqMan™ Genotyping Assays and Instruments:
- TaqMan™ Genotyping Master Mix has been tested across all types of TaqMan™ SNP Genotyping Assays: TaqMan™ Drug Metabolism Genotyping Assays, TaqMan™ SNP Genotyping Assays, and Custom TaqMan™ SNP Genotyping Assays
- In addition, the master mix is validated with the Applied Biosystems™ thermal cyclers and real-time PCR systems
High-Throughput Setup and Analysis:
- The combination of several low-cost thermal cyclers for PCR and a single real-time PCR instrument for allelic discrimination make high throughput SNP genotyping manageable
- TaqMan™ Genotyping Master Mix further improves high-throughput setup and analysis with excellent room temperature stability both before and after PCR
- This provides the flexibility required to run long experiments, unattended overnight or over a weekend
Reliable Discrimination with Challenging Targets
- Amplicon design constraints make SNP detection challenging, particularly when targets contain an abundance of GC- or AT-rich regions
- The optimized components of TaqMan™ Genotyping Master Mix result in tight, well-separated clusters and more accurate allele-calls compared to other mixes, even for challenging targets
Setting the Industry Standard for Quality:
- Extensive analytical and functional tests are performed for each manufactured lot of TaqMan™ Genotyping Master Mix, including one TaqMan™ SNP Genotyping Assay and one TaqMan™ Drug Metabolism Genotyping Assay
- Results are compiled in an informative Certificate of Analysis, which is accessible through the Applied Biosystems™ Web site
Order Info
Guaranteed minimum shelf life is 60 days. Shipping Conditions: Wet Ice
Specifications
Specifications
| Concentration | 2X |
| Content And Storage | Supplied at 2X concentration. The mix is optimized for SNP genotyping using TaqMan™ probes and contains AmpliTaq Gold™ DNA Polymerase UP (Ultra Pure), dNTPs without dUTP, Passive Reference 1 and optimized mix components. This pack contains one 10 mL bottle, sufficient for 400 reactions at the 50 μL total volume. Storage: 2-8°C Guaranteed minimum shelf life is 60 days (exact expiry date printed on product and CofA). |
| Detection Method | Primer-probe |
| GC-Rich PCR Performance | High |
| PCR Method | qPCR |
| Polymerase | AmpliTaq Gold DNA Polymerase |
| Reaction Speed | Standard |
| Technique | SNP Genotyping (Real-Time PCR-Based), CNV (Copy Number Variation) |
| For Use With (Equipment) | Applied Biosystems StepOnePlus™ Fast Real-Time PCR System, QuantStudio™ 12k Flex, QuantStudio™ 3, QuantStudio™ 5, QuantStudio™ 6 Flex, QuantStudio™ 7 |
| Genotyping Target | Copy Number Variants, SNPs (Unknown or Numerous), Drug Metabolizing Enzymes, SNPs (Known) |
| Show More |
Frequently Asked Questions (FAQs)
We recommend using either the TaqPath ProAmp Master Mix or the TaqMan Genotyping Master Mix. The TaqMan Genotyping Master Mix has the advantage of proven performance with up to 3 days of pre- and post-PCR stability, allowing you to set up plates ahead of time or read the plates later (see the data here, https://tools.thermofisher.com/content/sfs/brochures/cms_039236.pdf) while the TaqPath ProAmp Master Mix can handle samples that may have inhibitors present.
If you are targeting a low-abundance gene, you may have trouble getting Ct values in a good, reliable range (Ct > 32). To increase the sensitivity of the assay, you may want to consider the following:
- Increase the amount of RNA input into your reverse transcription reaction, if possible
- Increase the amount of cDNA in your qPCR reaction (20% by volume max)
- Try a different reverse transcription kit, such as our SuperScript VILO Master Mix, for the highest cDNA yield possible
- Consider trying a one-step or Cells-to-CT type workflow (depending on your sample type)
Most times your instrument software can automatically set a proper baseline for your data. Check out our short video, Understanding Baselines, for more information on how to set them (https://www.youtube.com/watch?feature=player_embedded&v=5BjFAJHW-bE).
In most cases your instrument software can automatically set a proper threshold for your data. Check out our short video, Understanding Thresholds, for more information on how to set them (https://www.youtube.com/watch?feature=player_embedded&v=H_xsuRQIM9M).
There could be several reasons for no amplification from an assay or primer set. Please see these examples and suggested solutions in our Real-Time Troubleshooting Tool (https://www.thermofisher.com/us/en/home/life-science/pcr/real-time-pcr/qpcr-education/real-time-pcr-troubleshooting-tool/gene-expression-quantitation-troubleshooting/no-amplification.html) for more details.
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.