Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TAS1R2 Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB166165 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB166165 100 μg
NB166166 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB166165 Supplier Novus Biologicals Supplier No. NBP315535100UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TAS1R2 Polyclonal antibody specifically detects TAS1R2 in Human samples. It is validated for Western Blot

Specifications

Antigen TAS1R2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS, 50% glycerol, pH7.3
Gene Alias G protein-coupled receptor 71, Sweet taste receptor T1R2, T1R2GPR71, taste receptor type 1 member 2, taste receptor, type 1, member 2, TR2G-protein coupled receptor 71
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 450-550 of human TAS1R2 (NP_689418.2).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 80834
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.