Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TAS2R38 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$254.42 - $728.30
Specifications
| Antigen | TAS2R38 |
|---|---|
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
TAS2R38 Polyclonal specifically detects TAS2R38 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| TAS2R38 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| P59533 | |
| 5726 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: NAKLRRAVMTILLWAQSSLKVRADHKADSRTL | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Polyclonal | |
| Rabbit | |
| GPCR | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| PTCphenylthiocarbamide tasting, T2R38, T2R61PTC bitter taste receptor, taste receptor type 2 member 38, Taste receptor type 2 member 61, taste receptor, type 2, member 38 | |
| TAS2R38 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title