Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TAS2R4 Antibody, Novus Biologicals™
SDP

Catalog No. NB362835 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB362835 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB362835 Supplier Novus Biologicals Supplier No. NBP130553
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TAS2R4 Polyclonal antibody specifically detects TAS2R4 in Human samples. It is validated for Western Blot, Immunohistochemistry

Specifications

Antigen TAS2R4
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry
Formulation No buffer
Gene Alias candidate taste receptor T2R4, taste receptor, type 2, member 4
Host Species Rabbit
Immunogen A synthetic peptide from human TAS2R4 conjugated to an immunogenic carrier protein was used as the immunogen.
Purification Method Ammonium sulfate precipitation
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 50832
Reconstitution Reconstitute in 500 ul of sterile water. Centrifuge to remove any insoluble material. When reconstituting, glycerol (1:1) may be added for additional stability for storage
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.