Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TAS2R9 Antibody, Novus Biologicals™
SDP

Catalog No. NB11074894 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB11074894 0.1 mL
NB018015 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB11074894 Supplier Novus Biologicals Supplier No. NB11074894
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TAS2R9 Polyclonal specifically detects TAS2R9 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen TAS2R9
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:300-1:2000, Immunohistochemistry 1:300-1:2000, Immunohistochemistry-Paraffin 1:300-1:2000, Immunohistochemistry-Frozen 1:300-1:2000
Formulation Lyophilized from whole antisera with No Preservative
Gene Accession No. Q9NYW1
Gene Alias T2R9TRB6Taste receptor family B member 6, taste receptor type 2 member 9, taste receptor, family B, member 6, taste receptor, type 2, member 9
Gene Symbols TAS2R9
Host Species Rabbit
Immunogen A synthetic peptide as a part of human taste receptor, type 2, member 9 (TAS2R9, TA2R9, T2R9, TRB6) conjugated to immunogenic carrier protein was used as the immunogen.
Purification Method Unpurified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 50835
Test Specificity TAS2R9.
Reconstitution Reconstitute in 0.1 ml of sterile water. Centrifuge to remove any insoluble material. Glycerol may be added (1:1) for additional stability. Please note the sample size is provided in reconstituted format.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.