Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Tasimelteon
GREENER_CHOICE

Catalog No. p-7250115
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective dual melatonin receptor (MT1/MT2) agonist

Tasimelteon is a high affinity and selective dual melatonin receptor (MT1/MT2) agonist (Ki values are 0.304 nM and 0.0692 nM, respectively). It has no significant interaction with any other commonly screened receptors or enzyme binding sites. Tasimelteon induces a potent, concentration-dependent inhibition of forskolin-stimulated cAMP accumulation in NIH-3T3 cells stably expressing human MT1 (EC50 = 0.74 nM) or expressing human MT2 (EC50 = 0.1 nM) receptors. Tasimelteon is a circadian regulator.

Specifications

Chemical Name or Material N-[[(1 R,2 R)-2-(2,3-Dihydro-4-benzofuranyl)cyclopropyl]methyl]propanamide
CAS 609799-22-6
Quantity 10 mg
Target Melatonin (MT) Receptor Agonists
Molecular Formula C15H19NO2
Purity 0.98
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.