Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TASP1 Antibody (OTI1H6), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NP274444A30 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP274444A30 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP274444A30 Supplier Novus Biologicals Supplier No. NBP274444AF350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TASP1 Monoclonal specifically detects TASP1 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen TASP1
Applications Western Blot
Classification Monoclonal
Clone OTI1H6
Conjugate Alexa Fluor 350
Dilution Western Blot
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias C20orf13taspase 1, chromosome 20 open reading frame 13, dJ585I14.2, EC 3.4.25.-, FLJ20212, MGC39159, taspase, threonine aspartase, 1, Taspase-1, threonine aspartase 1
Gene Symbols TASP1
Host Species Mouse
Immunogen Full length recombinant protein of human TASP1 (NP_060184) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 55617.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.