Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TAT Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112736
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112736 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112736 Supplier Invitrogen™ Supplier No. PA5112736
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TAT Polyclonal Antibody for ELISA, ICC/IF, IHC (P), Western Blot

TAT is a mitochondrial protein tyrosine aminotransferase which is present in the liver and catalyzes the conversion of L-tyrosine into p-hydroxyphenylpyruvate.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TAT
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene TAT
Gene Accession No. P17735
Gene Alias 2.6.1.5; L-tyrosine:2-oxoglutarate aminotransferase; Tat; testis tissue sperm-binding protein Li 34a; tyrosine aminotransferase; tyrosine aminotransferase, cytosolic; tyrosine transaminase
Gene Symbols TAT
Host Species Rabbit
Immunogen Recombinant Human Tyrosine aminotransferase protein (25-111AA).
Purification Method Protein G, Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6898
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.